Maleic hydrazide ≥97%
Supplier: BeanTown Chemical
123810-100G
123810-500G
BT123810-100GEA
46
USD
BT123810-100G
BT123810-500G
Maleic hydrazide ≥97%
Maleic hydrazide
CAS: 123-33-1; EC No: 204-619-9; MDL No: MFCD00006665; RTECS: UR5950000
Powder; Molecular Formula: C4H4N2O2 ; MW: 112.09
Melting Point: 299-301° (decomposes); Flash point: 300°C (572°F)
- RTECS: UR5950000
- Powder
Formula:
C₄H₄N₂O₂ Melting Pt: 299…301 °C (dec.) Flash Pt: 300 °C (572 °F) |
MDL Number:
MFCD00006665 CAS Number: 123-33-1 EINECS: 204-619-9 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...