Dihydroartemisinin ≥98%

Supplier: BeanTown Chemical

123740-1G 123740-5G
BT123740-1GEA 98 USD
BT123740-1G BT123740-5G
Dihydroartemisinin ≥98%
Dihydroartemisinin

CAS: 71939-50-9; MDL No: MFCD00274495 Solid; Molecular Formula: C15H24O5; MW: 284.35 Melting Point: 144-149°; Boiling Point: 376° Density (g/mL): 1.24


  • Solid

Formula: C₁₅H₂₄O₅
Boiling Pt: 376 °C
Melting Pt: 144…149 °C
Density: 1.24 g/mL
MDL Number: MFCD00274495
CAS Number: 71939-50-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR