Tantalum(V) oxide ≥99.95%

Supplier: BeanTown Chemical

Synonyms: Ditantalum pentaoxide

123625-100G 123625-25G
BT123625-100GEA 642 USD
BT123625-100G BT123625-25G
Tantalum(V) oxide ≥99.95%
Tantalum (V) oxide

CAS: 1314-61-0; EC No: 215-238-2; MDL No: MFCD00011254 3-12mm Sintered Lump; Linear Formula: Ta2O5; MW: 441.89 Melting Point: 1800° Density (g/mL): 8.2


  • 3-12mm Sintered Lump

Formula: O₅Ta₂
Melting Pt: 1800 °C
Density: 8.2 g/mL
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR