Sodium chloride ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
123505-250G
123505-50G
BT123505-250GEA
225
USD
BT123505-250G
BT123505-50G
Sodium chloride ≥99.99% (trace metals basis)
Sodium chloride
CAS: 7647-14-5; EC No: 231-598-3; MDL No: MFCD00003477; RTECS: VZ4725000
Crystalline; Linear Formula: NaCl; MW: 58.44
Melting Point: 801°; Boiling Point: 1413°
Density (g/mL): 2.165; Refractive Index: 1.544
- RTECS: VZ4725000
- Refractive index: 1.544
- Crystalline
Formula:
NaCl Boiling Pt: 1413 °C Melting Pt: 801 °C Density: 2.165 g/mL |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...