2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine

Supplier: BeanTown Chemical

Synonyms: 2-((4-Chlorophenyl)(piperidin-4-yloxy)methyl)pyridine

123450-1G
BT123450-1GEA 606.58 USD
BT123450-1G
2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine
2-[(4-Chlorophenyl)(4-piperidinyloxy)methyl]pyridine

  • Solid

Formula: C₁₇H₁₉ClN₂O
MW: 302.802 g/mol
Density: 1.217 g/mL
MDL Number: MFCD13184723
CAS Number: 122368-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR