Cobalt ≥99.995% (trace metals basis), wire Ø 0.5 mm

Supplier: BeanTown Chemical

123435-2M 123435-50CM
BT123435-2MEA 437.14 USD
BT123435-2M BT123435-50CM
Cobalt ≥99.995% (trace metals basis), wire Ø 0.5 mm
Cobalt

CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000 Solid; Molecular Formula: Co; MW: 58.93 Melting Point: 1495°; Boiling Point: 2900° Density (g/mL): 8.92


  • RTECS: GF8750000
  • Solid

Formula: Co
Boiling Pt: 2900 °C
Melting Pt: 1495 °C
Density: 8.92 g/mL
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR