5-Bromo-2,4-dichloro-6-methylpyrimidine ≥95%

Supplier: BeanTown Chemical

123305-1G 123305-5G
BT123305-1GEA 92 USD
BT123305-1G BT123305-5G
5-Bromo-2,4-dichloro-6-methylpyrimidine ≥95%
5-Bromo-2,4-dichloro-6-methylpyrimidine

CAS: 56745-01-8 Solid; Molecular Formula: C5H3BrCl2N2; MW: 241.90


  • Solid

Formula: C₅H₃BrCl₂N₂
MW: 241.9000 g/mol
MDL Number: MFCD00504524
CAS Number: 56745-01-8

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR