1-Pentanesulfonic acid sodium salt, (max. 1% H₂O) ≥99% (dry weight)
Supplier: BeanTown Chemical
122695-100G
122695-25G
122695-5G
BT122695-100GEA
367
USD
BT122695-100G
BT122695-25G
BT122695-5G
1-Pentanesulfonic acid sodium salt, (max. 1% H₂O) ≥99% (dry weight)
1-Pentanesulphonic acid sodium salt
CAS: 22767-49-3; EC No: 245-208-4; MDL No: MFCD00007541
Solid; Linear Formula: CH3(CH2)4SO3Na; Molecular Formula: C5H11NaO3S; MW: 174.19
Melting Point:
- Solid
Formula:
C₅H₁₁O₃SNa Melting Pt: >300 °C |
MDL Number:
MFCD00007541 CAS Number: 22767-49-3 EINECS: 245-208-4 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...