4-Methyl-1,10-phenanthroline
Supplier: BeanTown Chemical
122355-1G
122355-5G
BT122355-1GEA
86
USD
BT122355-1G
BT122355-5G
4-Methyl-1,10-phenanthroline
4-Methyl-1,10-phenanthroline
CAS: 31301-28-7; MDL No: MFCD00004978
Crystalline; Molecular Formula: C13H10N2; MW: 194.23
Melting Point: 143-145°
- Crystalline
Formula:
C₁₃H₁₀N₂ Melting Pt: 143…145 °C |
MDL Number:
MFCD00004978 CAS Number: 31301-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...