Bromophenol blue sodium salt ACS

Supplier: BeanTown Chemical

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulfonate

122340-100G 122340-25G 122340-5G
BT122340-100GEA 215.02 USD
BT122340-100G BT122340-25G BT122340-5G
Bromophenol blue sodium salt ACS
Bromophenol blue sodium salt

CAS: 34725-61-6; EC No: 252-170-2; MDL No: MFCD00013793 Crystalline; Molecular Formula: C19H9Br4NaO5S; MW: 691.94


  • Crystalline

Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR