6-Benzylaminopurine ≥99%

Supplier: BeanTown Chemical

122330-1G 122330-25G 122330-5G
BT122330-1GEA 24.23 USD
BT122330-1G BT122330-25G BT122330-5G
6-Benzylaminopurine ≥99%
6-Benzylaminopurine

CAS: 1214-39-7; EC No: 214-927-5; MDL No: MFCD00005572; RTECS: UD3150000 Crystalline/Powder; Molecular Formula: C12H11N5; MW: 225.25 Melting Point: 230-233°


  • RTECS: UD3150000
  • Crystalline/Powder

Formula: C₁₂H₁₁N₅
Melting Pt: 230…233 °C
MDL Number: MFCD00005572
CAS Number: 1214-39-7
EINECS: 214-927-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR