4-Iodo-3-nitroanisole 98

Supplier: BeanTown Chemical

Synonyms: 1-Iodo-4-methoxy-2-nitrobenzene

122185-25G 122185-5G
BT122185-25GEA 245 USD
BT122185-25G BT122185-5G
4-Iodo-3-nitroanisole 98
4-Iodo-3-nitroanisole

CAS: 58755-70-7; MDL No: MFCD00024195 Solid; Molecular Formula: C7H6INO3; MW: 279.04 Melting Point: 60-63° Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₇H₆INO₃
MW: 279.03 g/mol
Melting Pt: 60…63 °C
MDL Number: MFCD00024195
CAS Number: 58755-70-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR