Quinaldine ≥97%
Supplier: BeanTown Chemical
Synonyms:
2-Methylquinoline
121920-100G
121920-25G
121920-500G
BT121920-100GEA
66
USD
BT121920-100G
BT121920-25G
BT121920-500G
Quinaldine ≥97%
Quinaldine
CAS: 91-63-4; EC No: 202-085-1; MDL No: MFCD00006756; RTECS: UZ9625000
Liquid; Molecular Formula: C10H9N ; MW: 143.19
Melting Point: -2°; Boiling Point: 248°; Flash point: 79°C (174°F)
Density (g/mL): 1.058; Refractive Index: 1.612
Air Sensitive, Light Sensitive, Moisture Sensitive
- RTECS: UZ9625000
- Refractive index: 1.612
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 247.6 Melting Pt: -2 Density: 1.058 g/mL Flash Pt: 79 °C (174 °F) |
MDL Number:
MFCD00006756 CAS Number: 91-63-4 EINECS: 202-085-1 Merck Index: 12,08228 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...