Cobalt ≥99.995% (trace metals basis), rods Ø 12 mm
Supplier: BeanTown Chemical
121895-50MM
121895-10MM
BT121895-50MMEA
1243.34
USD
BT121895-50MM
BT121895-10MM
Cobalt ≥99.995% (trace metals basis), rods Ø 12 mm
Cobalt
- RTECS: GF8750000
- Solid
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2900 °C Melting Pt: 1495 °C Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...