(R)-(+)-α,α-Diphenylprolinol ≥99%

Supplier: BeanTown Chemical

Synonyms: (R)-(+)-alpha,alpha-Diphenylprolinol, (R)-(+)-α,α-Diphenyl-2-pyrrolidinemethanol, (R)-(+)-2-(Diphenylhydroxymethyl)pyrrolidine, α,α-Diphenyl-D-prolinol

121830-1G 121830-250MG
BT121830-1GEA 160.79 USD
BT121830-1G BT121830-250MG
(R)-(+)-α,α-Diphenylprolinol ≥99%
(R)-(+)-α,α-Diphenylprolinol

CAS: 22348-32-9; MDL No: MFCD00077754 Crystalline; Molecular Formula: C17H19NO; MW: 253.34 Melting Point: 77-80° Optical Rotation: [α]20/D +69°, c = 3 in chloroform


  • Optical rotation: [α]20/D +69°, c = 3 in chloroform
  • Crystalline

Formula: C₁₇H₁₉NO
Melting Pt: 77…80 °C
MDL Number: MFCD00077754
CAS Number: 22348-32-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR