Magnesium fluoride ≥99.9% (trace metals basis excluding Ca, Na)
Supplier: BeanTown Chemical
121675-100G
121675-25G
BT121675-100GEA
197
USD
BT121675-100G
BT121675-25G
Magnesium fluoride ≥99.9% (trace metals basis excluding Ca, Na)
Magnesium fluoride
CAS: 7783-40-6; EC No: 231-995-1; MDL No: MFCD00011108; RTECS: OM3325000
3-6mm Vacuum Melted Lump; Linear Formula: MgF2; MW: 62.30
Melting Point: 1621°
Density (g/mL): 3.148
- RTECS: OM3325000
- 3-6mm Vacuum Melted Lump
Formula:
MgF₂ MW: 62.3 g/mol Boiling Pt: 2260 Melting Pt: 1266 Density: 3.148 g/mL |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 12,05701 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...