2-Ethynylpyridine ≥98% stabilised
Supplier: BeanTown Chemical
121040-1G
121040-25G
121040-5G
BT121040-1GEA
44
USD
BT121040-1G
BT121040-25G
BT121040-5G
2-Ethynylpyridine ≥98% stabilised
2-Ethynylpyridine
CAS: 1945-84-2; MDL No: MFCD00041598
Liquid; Molecular Formula: C7H5N; MW: 103.12
Boiling Point: 85°/12 mmHg; Flash point: 76°C (168°F)
Density (g/mL): 1.021; Refractive Index: 1.56
- Refractive index: 1.56
- Liquid
Formula:
C₇H₅N Boiling Pt: 85 °C/12 mmHg Density: 1.021 g/mL Flash Pt: 76 °C (168 °F) |
MDL Number:
MFCD00041598 CAS Number: 1945-84-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...