Sulfo-SANPAH (Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate)

Synonyms: Sulfo SANPAH

10189-582EA 604.44 USD
10189-582 10189-584
Sulfo-SANPAH (Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate)
Sulfo-SANPAH (Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate)
Formula: C₁₆H₁₇O₉N₆NaS
MW: 492.40 g/mol
CAS Number: 102568-43-4

Order Now

Specification Test Results

CAS 102568-43-4
Purity ≥90% by NMR
Form Red orange solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR