Tween® 20 (Polysorbate) 10% in aqueous solution, MegaPure™ detergent

10191-298EA 181.62 USD
10191-298 10191-300 10191-302 76290-156
Tween® 20 (Polysorbate) 10% in aqueous solution, MegaPure™ detergent
Tween® 20 (Polysorbate)

A non-ionic detergent useful for the isolation of membrane-bound proteins under non-denaturing conditions.


  • A non-ionic detergent

Formula: C₅₈H₁₁₄O₂₆
MDL Number: MFCD00165986
CAS Number: 9005-64-5

Order Now

Specification Test Results

CAS 9005-64-5
Purity Aldehydes: 200 µM; Peroxides: 200 µM
Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR