Blue Epoxy-Coated Steel Wire Test Tube Racks
Supplier: Wards
CH182015A CH182015E CH182015F CH182015G CH182015H CH182015B CH182015C CH182015D
470165-390EA 29.9 USD
470165-390 470165-442 470165-444 470165-446 470165-448 470165-452 470165-454 470165-456
Blue Epoxy-Coated Steel Wire Test Tube Racks
Racks Tube Racks
Coated steel wire racks are chemical and corrosion resistant and come in a variety of sizes.
  • Test tube rack featuring a sturdy epoxy-coated steel construction that will stand up to the wear and tear any lab
  • Withstands temperatures up to 121 °C (249.8 °F)
Coating provides a protective and smooth coat that is impervious to weak acids, bases, and salts.