Sterile Cotton Swabs, Texwipe®
Supplier: Texwipe
STX705W STX705P
10127-574BX 32.82 USD
10127-574 10127-576
Sterile Cotton Swabs, Texwipe®
Swabs Swabs for Cleaning
Sterile cotton swabs are constructed from 100% USP grade cotton. These swabs are bonded securely to a wooden or polystyrene shaft by an aqueous-based adhesive.
  • Gamma irradiated
  • Pre-sterilized swabs
  • Packaging labeled for global use
  • Widely used in pharmaceutical and microbiological Laboratories
  • Certificates of irradiation and compliance attached to each case
The swabs are made to exacting and consistent tolerances using high precision automated processes. The easy to open, individually wrapped swabs ensure sterility at point-of-use. The swabs are visible through clear packaging which also offers visibility of seal integrity.