(±)-2-Aminoheptane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2-Aminoheptane, (±)-Tuaminoheptane, 1-Methylhexylamine, 2-Heptylamine, (±)-1-Methylhexylamine, Tuaminoheptane

M-Clarity™ quality level = MQ200

8.41027.0025 8.41027.0100 8.41027.0010
EM8.41027.0025CS 4864 USD
EM8.41027.0025 EM8.41027.0100 EM8.41027.0010
(±)-2-Aminoheptane for synthesis, Sigma-Aldrich®
(±)-2-Aminoheptane
Formula: C₇H₁₇N
MW: 115.22 g/mol
Boiling Pt: 143
MDL Number: MFCD00008101
CAS Number: 123-82-0
UN: 2733
ADR: 3 (8) III
Merck Index: 12,09934

Order Now

Specification Test Results

CAS 123-82-0
Synonyms 2-Heptylamine, 1-Methylhexylamine
Hill Formula C₇H₁₇N
Molar Mass 115.22 g/mol
Boiling point 142 - 144 °C
Density 0.768 g/cm³ (20 °C)
Flash point 54 °C
pH value (10 g/l, H₂O, 20 °C)
Assay (GC, area%) ≥ 98.0 % (a/a)
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR