Oscilloscope Trainer
Supplier: AMERICAN 3B SCIENTIFIC SE
1000902
470162-782EA 2063.49 USD
470162-782
Oscilloscope Trainer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Investigate the design and operation of modern cathode ray tubes.
  • Device comes with a socket and printed wiring scheme
The electron beam can be deflected by an electric field produced by the deflection plates integrated into the tube, and by a magnetic field from three external coils mounted on a ring. A Wehnelt cylinder is used to focus the beam. A gas filling and fluorescent screen makes it possible to observe the beam in the tube. A continuously adjustable saw-tooth generator can be used to analyze and visualize time dependent processes.