Horse Serum, Vector Laboratories
Supplier: Vector Laboratories
S-2000-20
101098-384EA 182.77 USD
101098-384
Horse Serum, Vector Laboratories
Sera (Serum)
Normal Horse serum can be used as a blocking reagent in Immunoassays.
  • Used as a blocking reagent
  • Collected from healthy adult animals
  • Heat-treated and centrifuged
It is a pooled sample collected from healthy adult animals. The serum is heat-treated and centrifuged to remove precipitates and then filtered. Each serum is tested with the appropriate antibody to check for possible cross-reactivities. The sera are supplied undiluted with 0.08% sodium azide as a preservative.