Cobalt Glass Plates, United Scientific Supplies
Supplier: United Scientific Supplies
CGP2X2 CGP3X3 CGP4X4
470164-032PK 39.6 USD
470164-032 470164-034 470164-036
Cobalt Glass Plates, United Scientific Supplies
Filters Optical Filters
Improves visibility in sodium and potassium flame tests.
  • Uniform in color
  • Available in various sizes
High-quality dark blue cobalt glass filter is used in sodium and potassium salt flame tests. When a flame is viewed through this filter, the color orange is blocked so that a violet-colored flame is visible.