EZ-SPORE™ Quantitative QC Microorganism Preparations, Microbiologics
Supplier: MICROBIOLOGICS, INC.
Food microbiology labs rely on EZ-SPORE™ quantitative QC microorganism preparations to support their quality assurance programs as daily process controls for spoilage detection methods
Packaged in a vial containing 10 pellets of a single lyophilized QC microorganism. Saves time and reduces labor by delivering a specific number of colony forming units (CFU).
EZ-SPORE™ preparations provide a guaranteed concentration of 104 CFU per pellet.
Certifications: Preparations come with complete and convenient peel-off Certificate of Assay documentation. Assured quality, traceable to reference culture.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.