Axygen® Automation Tips for Molecular Devices FlexStation®, Corning
Supplier: Corning
Axygen®
RMT-200-BK-R RMT-200-BK-R
10770-594CS 483.74 USD
10770-594 CA10770-594
Axygen® Automation Tips for Molecular Devices FlexStation®, Corning
Pipette Tips
Robbins-style tips for Beckman Multimek 'Type II' systems.
  • Accuracy and precision testing is performed for every manufacturing lot using the finest automated systems in the industry
  • Certified RNase-, DNase-, and endotoxin-free
  • Available in black or transparent
  • Non filtered, non sterile
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. Available in nonsterile, sterile, non-filtered, and filtered formats. Black and clear polypropylene tips are available in 384-well format designed for Molecular Devices Flexstation systems.