Sodium dihydrogen phosphate monohydrate USP, FCC, Ph. Eur.
Supplier: Macco Organiques S.R.O
Synonyms:
Sodium dihydrogen orthophosphate 1-hydrate
51115010120
89523-446EA
896.34
USD
89523-446
Sodium dihydrogen phosphate monohydrate USP, FCC, Ph. Eur.
Sodium dihydrogen phosphate monohydrate
For excipient use only.
Certifications: Certificates: HACCP, ISO 9001:2008, Kosher including for Passover days, HALAL.
Ordering information: Minimum order quantities may apply; contact your VWR representative for more information.
Formula:
NaH₂PO₄·H₂O MW: 137.99 g/mol |
MDL Number:
MFCD00149208 CAS Number: 10049-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...