Anti-CDH2 Mouse Monoclonal Antibody [clone: 1D8B3]

Supplier: Proteintech
66219-1-IG-150UL
10500-700EA 462.47 USD
10500-700
Anti-CDH2 Mouse Monoclonal Antibody [clone: 1D8B3]
Antibodies
The N-cadherin antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human N-cadherin. This antibody recognizes human, pig, mouse, rat antigen. The N-cadherin antibody has been validated for the following applications: ELISA, IHC, WB analysis.

Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 1D8B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR