Anti-TFAP2E Rabbit Polyclonal Antibody

Supplier: Proteintech
25829-1-AP-150UL
10220-886EA 462.47 USD
10220-886
Anti-TFAP2E Rabbit Polyclonal Antibody
Antibodies
The TFAP2E antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TFAP2E. This antibody recognizes human, mouse, rat antigen. The TFAP2E antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: TFAP2E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR