Anti-MRPS5 Rabbit Polyclonal Antibody

Supplier: Proteintech
16428-1-AP-150UL
10222-102EA 462.47 USD
10222-102
Anti-MRPS5 Rabbit Polyclonal Antibody
Antibodies
The MRPS5 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MRPS5. This antibody recognizes human,mouse,rat antigen. The MRPS5 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: MRPS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR