Anti-GOD Rabbit Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-4637S 200-4637S
RL2004637SEA 148 USD
CA11029-052 RL2004637S
Anti-GOD Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: GOD
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Aspergillus niger
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR