Anti-Uricase Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-101-092-0100 200101092-0100
RL200101092010EA 518 USD
RL200101092010 CARL200101092
Anti-Uricase Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Uricase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bacillus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR