Hydrogen Peroxide WFI Sterile Solutions, STERIS®
Supplier: Steris
1S0708WR 1S0496 1S0796WR 1S0796WR 1S0508WR 1S0596WR 1S0408 1S0777WR 1S0777WR 1S0577WR 1S0708WR 1S0477
10755-294CS 774.03 USD
CA75780-030 10755-294 10755-296 10770-532 CA75780-044 10755-298 CA75780-034 10770-534 CA75780-042 CA75780-032 CA75780-046 10770-530
Hydrogen Peroxide WFI Sterile Solutions, STERIS®
Cleaning Agents Detergents and Surface Cleansers
STERIS® Hydrogen Peroxide WFI Sterile Solutions are manufactured with United States Pharmcopeia quality Water For Injection (WFI) and Hydrogen Peroxide
Sterile filtered and filled in a ISO Class 5 aseptic environment, each package is double bagged for easy entry into controlled environments and aseptic core areas. These products have an extensive documentation package to meet validation objectives and are supported by the STERIS® Technical Services team for individualized application and validation assistance.
This low residue cleaner with minimal organic carbon is designed to remove organic and inorganic residues from product contact surfaces, room surfaces, equipment, gloves, etc.
Certifications: Solution is manufactured in an FDA cGMP, ISO 9001 and ISO 13485 Compliant Quality System environment.