HyperSep™ Retain SPE CX Cartridges, Thermo Scientific
Supplier: Thermo Scientific
HyperSep™ Retain™
60107-315 60107-307 60107-314 60107-308 60107-303 60107-304 60107-312 60107-311 60107-306 60107-305 60107-301
10047-196EA 413.05 USD
10047-196 10047-208 10047-218 10047-204 10047-214 10047-206 10047-216 10047-200 10047-210 10047-202 10047-212
HyperSep™ Retain SPE CX Cartridges, Thermo Scientific
Chromatography Columns
Achieve consistent, high recoveries for basic compounds with these Thermo Scientific HyperSep Retain CX Cartridges, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with sulfonic acid functional groups to give superior recoveries for basic analytes.
  • High and consistent recoveries
  • Fast and easy sample preparation and method development
  • pH stable zero to 14
Widely used in forensic and clinical analysis of a wide range of drugs of abuse, including basic and neutral drugs. HyperSep Retain SPE cartridges conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation. High, consistent recoveries. Ideal for large samples. Fast and easy sample preparation and method development. Reproducible, reliable sample preparation in a traditional format. pH stable 0 to 14. Available in a wide range of chemistries and bed weights. For use with the Thermo Scientific HyperSep-96 System.
Used in SPE sample preparation, these cation exchange cartridges can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.