National LoVial™ 13 mm Glass Crimp/Snap-Ring Vials, Thermo Scientific
Supplier: Thermo Scientific
C4015-4
66065-384CS 491.13 USD
66065-384
National LoVial™ 13 mm Glass Crimp/Snap-Ring Vials, Thermo Scientific
Vials
Manufactured from superior quality 33 expansion borosilicate clear Type 1, Class A glass, Thermo Scientific National 13 mm Snap/Crimp Vials feature a 13 mm crimp/snap-ring finish for use with 13 mm Aluminum Seals or Kim-Snap Closures.
See the Columns and Consumables Catalog for an autosampler compatibility list.
Polyspring inserts are self-aligning and provide a cushion against needle contact.
Delivery information: Autosampler compatibility includes Dionex, Shimadzu, Spark Holland, Varian, VWR (Merck)/Hitachi, and Waters (Wisp 48 Position Carousel).