Epower™ Certified Reference Material (CRM), Microbiologics
Supplier: MICROBIOLOGICS, INC.
New Epower Certified Reference Material (CRM) is a quantitative QC microorganism preparation designed for use in ISO 17025 Accredited Laboratories. For Testing Laboratories, Section 5.6.3.2 of ISO 17025:2005 states “Reference materials shall, where possible, be traceable to SI units of measurement, or to certified reference materials”. Epower CRM is available in concentrations ranging from 102 to 108 CFU per pellet.
Epower Certified Reference Material (CRM) includes a vial of 10 lyophilized quantitative QC microorganism pellets, instructions for use and a peel-off identification label with Certified Value for easy documentation.
Comprehensive Certificate of Analysis documents each lot's Certified Value, Expanded Uncertainty and Standard Deviation. Does not require frozen storage conditions. Easy storage, with no dry ice required, reduces shipping costs.
Certifications: 100% Traceable. Sourced from licensed, traceable strains. One passage from reference culture.
Ordering information: To receive this product, customers must have a valid ATCC End User Licensing Agreement in place with Microbiologics. Registration for this agreement is only required once, per location during the first order. Instructions for how to register can be downloaded from the website. Go to the "Related Information" box, click on "Additional Product Details", then "Instructions for End User Registration with Microbiologics".
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC® cultures.