Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Supplier: Thermo Scientific
C4000-LV1 C4000-1 C5000-680
66030-996PK 309.99 USD
66030-996 66030-998 66031-045 66064-858 66064-910 66064-912 66064-914 66064-952 89187-282 97054-232 97054-234 97054-240 97054-242 66030-000 66030-006 66030-990 66030-992 66030-994 89235-910 89235-912 89235-918 82428-552 10766-770 10781-888
Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Vials
Nearly eliminate septa push-through and improve sealing with Advanced Vial Closure System technology. This innovation not only prevents septa push-through, but provides a stop point for the cap to prevent over - or under-tightening and provide an optimal, consistent seal every time. Vials are made from superior quality 33 expansion borosilicate clear Type 1, Class A glass, and meet all US, EU, and JPN Pharmacopia standards.
  • Closure system prevents septa push-through
  • Wide neck opening
Fill vials easily with the wide neck opening; requires 6 mm diameterMore Product Information