Boc-D-Ser-OBzl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Boc-D-serine benzyl ester

H66514-03 H66514-06 H66514-MD
AAH66514-03EA 56.96 USD
AAH66514-03 AAH66514-06 AAH66514-MD
Boc-D-Ser-OBzl 95%
Boc-D-Ser-OBzl
Formula: C₁₅H₂₁NO₅
Melting Pt: 66…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080265
CAS Number: 141527-78-8

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR