2-Amino-2'-chloro-5-nitrobenzophenone 98%
Supplier: Thermo Scientific Chemicals
H66440-06
H66440-14
H66440-22
AAH66440-06EA
34.3
USD
AAH66440-06
AAH66440-14
AAH66440-22
2-Amino-2'-chloro-5-nitrobenzophenone 98%
2-Amino-2'-chloro-5-nitrobenzophenone
2-Amino-2'-chloro-5-nitrobenzophenone, 98%
Purity: 98%
CAS No.: 2011-66-7
Molecular Formula: C13H9ClN2O3
Formula Weight: 276.68
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Pale yellow to yellow
Melting Point: 119-123°
Beilstein Reference: 2814216
MDL No.: MFCD00792453
Formula:
C₁₃H₉ClN₂O₃ Melting Pt: 119…123 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00792453 CAS Number: 2011-66-7 EINECS: 217-929-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...