(S)-(-)-Tetrahydro-3-furoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66392-03
H66392-MD
AAH66392-03EA
803.99
USD
AAH66392-03
AAH66392-MD
(S)-(-)-Tetrahydro-3-furoic acid 97%
(S)-(-)-Tetrahydro-3-furoic acid
(S)-(-)-Tetrahydro-3-furoic acid, 97%
Purity: 97%
CAS No.: 168395-26-4
Molecular Formula: C5H8O3
Formula Weight: 116.12
Physical Form: Liquid
Appearance: Yellow to brown
Beilstein Reference: 1281203
Optical Rotation: -15° (c=0.1 in ethanol)
MDL No.: MFCD07369984
Formula:
C₅H₈O₃ Storage Temperature: Ambient |
MDL Number:
MFCD07369984 CAS Number: 168395-26-4 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...