(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 98% HPLC_ASSAY_METHOD
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-N-FMOC-Octylglycine, N-Fmoc-2-octyl-L-glycine, Fmoc-octyl-Gly-OH
H66031-03
H66031-06
AAH66031-03EA
93.9
USD
AAH66031-03
AAH66031-06
AAH66031-MD
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 98% HPLC_ASSAY_METHOD
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
N-Fmoc-2-octyl-L-glycine, 98%
Purity: 98%
CAS No.: 193885-59-5
Molecular Formula: C25H31NO4
Formula Weight: 409.53
Physical Form: Powder
Appearance: White
Melting Point: 140-143°
MDL No.: MFCD01311753
Formula:
C₂₅H₃₁NO₄ MW: 409.526 g/mol Melting Pt: 140…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD01311753 CAS Number: 193885-59-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...