L-Serinamide hydrochloride 95%
Supplier: Thermo Scientific Chemicals
L-Serinamide hydrochloride, 95%
Purity: 95%
CAS No.: 65414-74-6
Molecular Formula: C3H8N2O2.HCl
Formula Weight: 140.57
Physical Form: Powder
Appearance: White
Melting Point: 185-188°
Beilstein Reference: 3911773
Optical Rotation: +14°(c=1 in water)
MDL No.: MFCD00067560
Formula:
C₃H₉ClN₂O₂ Melting Pt: 185…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067560 CAS Number: 65414-74-6 EINECS: 265-753-1 |
Specification Test Results
Assay Minimum | 95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...