N-Methylpyridin-3-amine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63602-03
H63602-06
H63602-MD
AAH63602-03EA
104.99
USD
AAH63602-03
AAH63602-06
AAH63602-MD
N-Methylpyridin-3-amine 95%
N-Methylpyridin-3-amine
3-(Methylamino)pyridine, 95%
Purity: 95%
CAS No.: 18364-47-1
Molecular Formula: C6H8N2
Formula Weight: 108.14
Physical Form: Liquid
Appearance: Yellow
UN No.: 2810
MDL No.: MFCD01418130
Formula:
C₆H₈N₂ Storage Temperature: Ambient |
MDL Number:
MFCD01418130 CAS Number: 18364-47-1 UN: 2810 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...