Bismuth ≥99.9999% (metals basis), rods, Puratronic®, Length 145 mm (5.71 in), Diameter 10 mm (0.39 in)
Supplier: Thermo Scientific Chemicals
Puratronic
047229-KF
47229-KJ
AA47229-KJEA
2570.39
USD
AA47229-KJ
AA47229-KF
Bismuth ≥99.9999% (metals basis), rods, Puratronic®, Length 145 mm (5.71 in), Diameter 10 mm (0.39 in)
Bismuth
Bismuth rod, 10mm (0.39in) dia x 145mm (5.71in) long, Puratronic|r, 99.9999% (metals basis). ^=7.5g/cm.
Formula:
Bi MW: 208.98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 |
Specification Test Results
Assay Minimum | 99.9999 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...