Cerium tribromide, ultra dry ≥99.99%, beads
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide
Ampuled under argon
047194-03
047194-06
047194-14
AA47194-03EA
74.3
USD
AA47194-03
AA47194-06
AA47194-14
Cerium tribromide, ultra dry ≥99.99%, beads
Cerium tribromide
Cerium(III) bromide, ultra dry, 99.99% (metals basis). -10 Mesh Beads, Ampouled under argon.
Purity: 99.99%
CAS No.: 14457-87-5
Molecular Formula: CeBr3
Formula Weight: 379.83
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.18
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 733°
Boiling Point: 1560°
MDL No.: MFCD00016004
Formula:
CeBr₃ MW: 379.83 g/mol Boiling Pt: 1560 °C Melting Pt: 732 Density: 5.18 Storage Temperature: Ambient |
MDL Number:
MFCD00016004 CAS Number: 14457-87-5 EINECS: 238-447-0 Merck Index: 14,1995 |
Specification Test Results
Assay Minimum | 99.99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...