Platinum ≥99.9% (metals basis), amorphous powder APS 2.0-8.0 micron

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

047145-MC 047145-06 047145-03
AA47145-MCEA 42.13 USD
AA47145-MC AA47145-06 AA47145-03
Platinum ≥99.9% (metals basis), amorphous powder APS 2.0-8.0 micron
Platinum

Platinum powder, amorphous, APS 2.0-8.0 micron, 99.9% (metals basis). S.A. 0.5-1.5m^2/g, Tap density 0.5-2.5g/cm^3.

Formula: Pt
MW: 195.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay Minimum 99.9

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR