Memphis Survivor™ Cut Resistant Gloves, MCR Safety
Supplier: MCR Safety
9348LD 9348MD 9348SD 9348XLD
80081-626EA 38.27 USD
80081-626 80081-628 80081-630 80081-632 80081-634
Memphis Survivor™ Cut Resistant Gloves, MCR Safety
Gloves
Memphis Survivor™ string knit gloves are used for general purpose work applications and can provide cut and abrasion resistance using high performance yarns.
  • These gloves are made of Kevlar® material
  • Protect the hands when working with sharp objects
Various material weights provide longer durability or additional dexterity where needed. The 9348 is a 13 gauge option that is lightweight and provides a CPPT cut score of 3.
Ordering information: These gloves are packaged by the each.