Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1), powder for electrophoresis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulphonate, Amido Black 10B, Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulfonate

J66798-14 J66798-22
AAJ66798-14EA 54.32 USD
AAJ66798-14 AAJ66798-22
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1), powder for electrophoresis
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)

Amido Black 10B, Electrophoresis Reagent.


Applications: Used to stain proteins in polyacrylamide gels.


Applications: Used to stain proteins in polyacrylamide gels.

Formula: C₂₂H₁₄N₆Na₂O₉S₂
Storage Temperature: Ambient
MDL Number: MFCD00004017
CAS Number: 1064-48-8
EINECS: 213-903-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR