L(+)-Ascorbic acid 99+%, powder for electrophoresis
Supplier: Thermo Scientific Chemicals
Synonyms:
Antiscorbic vitamin, Ascorbic Acid, L-Ascorbic acid, Vitamin C, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
J66291-18
J66291-22
AAJ66291-18EA
27.9
USD
AAJ66291-18
AAJ66291-22
L(+)-Ascorbic acid 99+%, powder for electrophoresis
L(+)-Ascorbic acid
L-(+)-Ascorbic acid, Electrophoresis Grade, 99+%
Purity: 99+%
CAS No.: 50-81-7
Molecular Formula: C6H8O6
Formula Weight: 176.12
Physical Form: Powder
Density: 1.65
Appearance: White
Melting Point: 190-193° dec.
Solubility: Soluble in water at 0.5 M
Beilstein Reference: 84272
Optical Rotation: +21° (c=10 in water)
MDL No.: MFCD00064328
Formula:
C₆H₈O₆ Melting Pt: 190…193 °C (dec.) Density: 1.65 Storage Temperature: Ambient |
MDL Number:
MFCD00064328 CAS Number: 50-81-7 EINECS: 200-066-2 Merck Index: 14,830 |
|
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...